Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAAAGGCTATTACATCAGAGACA[A/G]TGGCAACAATTCAGGTAATATAGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606461 | ||||||||||||||||||||
Literature Links: |
TGS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TGS1 - trimethylguanosine synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317902.1 | 325 | Intron | NP_001304831.1 | |||
NM_024831.7 | 325 | Missense Mutation | AAT,AGT | N,S 51 | NP_079107.6 | |
XM_005251328.2 | 325 | Intron | XP_005251385.1 | |||
XM_006716485.2 | 325 | Missense Mutation | AAT,AGT | N,S 51 | XP_006716548.1 | |
XM_017014004.1 | 325 | Missense Mutation | AAT,AGT | N,S 51 | XP_016869493.1 |
TMEM68 - transmembrane protein 68 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |