Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAGCCTCCGCAGCACAGGACGT[A/G]GCCAGCACCTGGGACTCGAACTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602747 | ||||||||||||||||||||
Literature Links: |
DUSP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUSP4 - dual specificity phosphatase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001394.6 | 4040 | Silent Mutation | GCC,GCT | A,A 336 | NP_001385.1 | |
NM_057158.3 | 4040 | Silent Mutation | GCC,GCT | A,A 245 | NP_476499.1 | |
XM_011544428.2 | 4040 | Silent Mutation | GCC,GCT | A,A 245 | XP_011542730.1 |