Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGAGCCCACGGACACCACCTCAA[C/T]GGTAAGTTTTATTTTGTTTTGTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610686 | ||||||||||||||||||||
Literature Links: |
UBXN2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBXN2B - UBX domain protein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077619.1 | 212 | Intron | NP_001071087.1 | |||
XM_005251165.2 | 212 | Intron | XP_005251222.1 | |||
XM_005251166.1 | 212 | Missense Mutation | CGG,TGG | R,W 63 | XP_005251223.1 |