Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATCAGTAAAAAGTGTTTTGGC[A/T]ACCTGCAGTGAAAGCACACACACTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609110 | ||||||||||||||||||||
Literature Links: |
FBXO43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO43 - F-box protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029860.3 | 1962 | Silent Mutation | GTA,GTT | V,V 627 | NP_001025031.2 | |
XM_011516987.2 | 1962 | Silent Mutation | GTA,GTT | V,V 670 | XP_011515289.1 | |
XM_011516989.2 | 1962 | Silent Mutation | GTA,GTT | V,V 636 | XP_011515291.1 | |
XM_011516990.1 | 1962 | Silent Mutation | GTA,GTT | V,V 636 | XP_011515292.1 | |
XM_011516992.2 | 1962 | Intron | XP_011515294.1 | |||
XM_017013358.1 | 1962 | Intron | XP_016868847.1 |