Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGCGCAGTGGGCGGGCACGCGT[C/G]GAGGACTTCTACACGGTGGAGACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615899 | ||||||||||||||||||||
Literature Links: |
OLFML2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OLFML2A - olfactomedin like 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282715.1 | 326 | Intron | NP_001269644.1 | |||
NM_182487.3 | 326 | Silent Mutation | GTC,GTG | V,V 71 | NP_872293.2 | |
XM_005251760.4 | 326 | Silent Mutation | GTC,GTG | V,V 71 | XP_005251817.1 | |
XM_006716989.2 | 326 | Silent Mutation | GTC,GTG | V,V 71 | XP_006717052.1 |