Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCAACTACTGCTCAGAGCTGTC[A/G]GATATGAAGATCATGTCCCAAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615955 MIM: 610615 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC183 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC183 - coiled-coil domain containing 183 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039374.4 | 675 | Silent Mutation | TCA,TCG | S,S 205 | NP_001034463.4 |
CCDC183-AS1 - CCDC183 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RABL6 - RAB, member RAS oncogene family-like 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM141 - transmembrane protein 141 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |