Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGCTTACCTGGACCTAAAATGA[A/G]GGTTCCACCTTGCTGAGCTGGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
AAED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AAED1 - AhpC/TSA antioxidant enzyme domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153698.1 | 574 | Missense Mutation | CTC,TTC | L,F 180 | NP_714542.1 | |
XM_005251783.3 | 574 | Intron | XP_005251840.1 | |||
XM_005251784.4 | 574 | Intron | XP_005251841.1 |