Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATCGGGTAGGATGAATGGGTAG[C/T]CCAGCATGCTGACGGTGAGCACCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C9orf24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C9orf24 - chromosome 9 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA1161 - KIAA1161 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020702.4 | 1801 | Missense Mutation | GAC,GGC | D,G 545 | NP_065753.2 | |
XM_011517966.2 | 1801 | Missense Mutation | GAC,GGC | D,G 545 | XP_011516268.1 | |
XM_017014930.1 | 1801 | Missense Mutation | GAC,GGC | D,G 545 | XP_016870419.1 |