Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGTGCCCTGAGCGTCTCCATGG[A/C]CTGCCTGAGCCCCTCGCAGCTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608360 | ||||||||||||||||||||
Literature Links: |
LRRC8A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC8A - leucine rich repeat containing 8 family member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHYHD1 - phytanoyl-CoA dioxygenase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100876.1 | 938 | Missense Mutation | GAC,GCC | D,A 2 | NP_001094346.1 | |
NM_001100877.1 | 938 | Missense Mutation | GAC,GCC | D,A 2 | NP_001094347.1 | |
NM_174933.3 | 938 | Missense Mutation | GAC,GCC | D,A 2 | NP_777593.2 |