Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTTACTGCGCTTGCTCCAGGGT[A/G]GGCTGACGGAGTGTCCCTGCAGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613747 MIM: 609787 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF24 - kinesin family member 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194313.2 | 2579 | Silent Mutation | CCC,CCT | P,P 1285 | NP_919289.2 | |
XM_005251451.2 | 2579 | Silent Mutation | CCC,CCT | P,P 924 | XP_005251508.1 | |
XM_006716764.2 | 2579 | Silent Mutation | CCC,CCT | P,P 827 | XP_006716827.1 | |
XM_011517861.1 | 2579 | Silent Mutation | CCC,CCT | P,P 1316 | XP_011516163.1 | |
XM_011517863.2 | 2579 | Silent Mutation | CCC,CCT | P,P 1285 | XP_011516165.1 | |
XM_017014697.1 | 2579 | Silent Mutation | CCC,CCT | P,P 1285 | XP_016870186.1 | |
XM_017014698.1 | 2579 | Silent Mutation | CCC,CCT | P,P 996 | XP_016870187.1 |
UBAP1 - ubiquitin associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |