Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTAGTAGGTTGTCTGAGACTTGAC[A/G]AACTCGTGGAGGCAGCGCAGGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615587 MIM: 609288 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP188 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUP188 - nucleoporin 188 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3GLB2 - SH3 domain containing GRB2 like endophilin B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287045.1 | 1023 | Silent Mutation | TTC,TTT | F,F 260 | NP_001273974.1 | |
NM_001287046.1 | 1023 | Silent Mutation | TTC,TTT | F,F 256 | NP_001273975.1 | |
NM_020145.3 | 1023 | Silent Mutation | TTC,TTT | F,F 256 | NP_064530.1 | |
XM_005252098.2 | 1023 | Silent Mutation | TTC,TTT | F,F 256 | XP_005252155.1 | |
XM_005252100.2 | 1023 | Silent Mutation | TTC,TTT | F,F 235 | XP_005252157.1 | |
XM_005252101.3 | 1023 | UTR 5 | XP_005252158.1 | |||
XM_006717188.2 | 1023 | Silent Mutation | TTC,TTT | F,F 284 | XP_006717251.1 | |
XM_006717189.2 | 1023 | Silent Mutation | TTC,TTT | F,F 280 | XP_006717252.1 | |
XM_006717190.1 | 1023 | Silent Mutation | TTC,TTT | F,F 284 | XP_006717253.1 | |
XM_006717191.1 | 1023 | Silent Mutation | TTC,TTT | F,F 260 | XP_006717254.1 |