Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTGGGTTCACCAGGCGCCACGT[A/T]TCAGACCTGGAGGTGCAAAACGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609092 | ||||||||||||||||||||
Literature Links: |
FBXO10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO10 - F-box protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012166.2 | 3069 | Missense Mutation | GAA,GAT | E,D 901 | NP_036298.2 | |
XM_005251439.4 | 3069 | Intron | XP_005251496.1 | |||
XM_006716754.3 | 3069 | Missense Mutation | GAA,GAT | E,D 426 | XP_006716817.1 | |
XM_017014618.1 | 3069 | Missense Mutation | GAA,GAT | E,D 959 | XP_016870107.1 | |
XM_017014619.1 | 3069 | Missense Mutation | GAA,GAT | E,D 901 | XP_016870108.1 |
POLR1E - RNA polymerase I subunit E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |