Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCCCGATGTGTTTCACGAGGTT[C/T]GGCTCCACTACATAGGCCCTCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM246 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM246 - transmembrane protein 246 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303107.1 | 1496 | Silent Mutation | NP_001290036.1 | |||
NM_001303108.1 | 1496 | Silent Mutation | NP_001290037.1 | |||
NM_032342.2 | 1496 | Silent Mutation | NP_115718.1 | |||
XM_017015206.1 | 1496 | Silent Mutation | XP_016870695.1 |
TMEM246-AS1 - TMEM246 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |