Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTTTTCTCAAGGGAAAAGTGT[C/T]AATCAGGTTGAACAGGGCCACAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300187 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRPX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRPX - sushi repeat containing protein, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170750.1 | 1459 | Missense Mutation | AAC,GAC | N,D 423 | NP_001164221.1 | |
NM_001170751.1 | 1459 | Missense Mutation | AAC,GAC | N,D 384 | NP_001164222.1 | |
NM_001170752.1 | 1459 | UTR 3 | NP_001164223.1 | |||
NM_006307.4 | 1459 | Missense Mutation | AAC,GAC | N,D 443 | NP_006298.1 | |
XM_017029893.1 | 1459 | Intron | XP_016885382.1 |