Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGGTATCTTTAAAAGAATCATC[C/T]TCATCTTCGCCATCTGAAACAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300687 MIM: 300252 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC6L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERCC6L - ERCC excision repair 6 like, spindle assembly checkpoint helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017669.2 | 3196 | Silent Mutation | GAA,GAG | E,E 1033 | NP_060139.2 |
PIN4 - peptidylprolyl cis/trans isomerase, NIMA-interacting 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170747.1 | 3196 | Intron | NP_001164218.1 | |||
NM_006223.3 | 3196 | Intron | NP_006214.2 |