Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTTCTTGATGCTCCTGGCATTG[A/T]TCTTCCGCTTTGTCAGATCTGTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300185 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP4 - A-kinase anchoring protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003886.2 | 2149 | Missense Mutation | GAA,GAT | E,D 675 | NP_003877.2 | |
NM_139289.1 | 2149 | Missense Mutation | GAA,GAT | E,D 666 | NP_647450.1 |