Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGACATGTAACGCACCTCCCCA[A/G]TTGATCCAGGGTAAGGCACGTAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300112 MIM: 300111 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGIX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGIX - MAGI family member, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099681.2 | 383 | Intron | NP_001093151.2 | |||
NM_001099682.2 | 383 | Intron | NP_001093152.2 | |||
NM_024859.3 | 383 | Silent Mutation | CAA,CAG | Q,Q 105 | NP_079135.3 | |
XM_005278062.4 | 383 | Intron | XP_005278119.1 | |||
XM_005278063.4 | 383 | Intron | XP_005278120.1 | |||
XM_005278064.4 | 383 | Silent Mutation | CAA,CAG | Q,Q 46 | XP_005278121.1 | |
XM_005278065.3 | 383 | Silent Mutation | CAA,CAG | Q,Q 46 | XP_005278122.1 | |
XM_006724560.3 | 383 | Silent Mutation | CAA,CAG | Q,Q 105 | XP_006724623.1 | |
XM_011543985.2 | 383 | Silent Mutation | CAA,CAG | Q,Q 46 | XP_011542287.1 | |
XM_011543986.2 | 383 | Silent Mutation | CAA,CAG | Q,Q 46 | XP_011542288.1 | |
XM_011543987.2 | 383 | Silent Mutation | CAA,CAG | Q,Q 46 | XP_011542289.1 |
PLP2 - proteolipid protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRICKLE3 - prickle planar cell polarity protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |