Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAAGCGACTGAGACCTGGCTTC[A/G]GCCCTTCTGACGTCATGTCTAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEND2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BEND2 - BEN domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184767.1 | 2356 | Intron | NP_001171696.1 | |||
NM_153346.4 | 2356 | Silent Mutation | GCC,GCT | A,A 768 | NP_699177.2 | |
XM_011545447.1 | 2356 | Silent Mutation | GCC,GCT | A,A 782 | XP_011543749.1 | |
XM_017029266.1 | 2356 | Silent Mutation | GCC,GCT | A,A 740 | XP_016884755.1 | |
XM_017029267.1 | 2356 | Intron | XP_016884756.1 |