Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCCATGGAGAATTCTTCACCAC[C/G]ATATTCCAGAGGAAGAATGTCTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600415 | ||||||||||||||||||||
Literature Links: |
TTPA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTPA - alpha tocopherol transfer protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000370.3 | 890 | Missense Mutation | CGT,GGT | R,G 246 | NP_000361.1 | |
XM_006716468.3 | 890 | Missense Mutation | CGT,GGT | R,G 130 | XP_006716531.1 |