Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGGCTCTCCTGGGGCAGAATAG[T/C]GTGCACCGCCAGGCTGCTGGGGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 190198 | ||||||||||||||||||||
Literature Links: |
NOTCH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NOTCH1 - notch 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017617.4 | 6655 | Missense Mutation | ACT,GCT | T,A 2455 | NP_060087.3 | |
XM_011518717.2 | 6655 | Missense Mutation | ACT,GCT | T,A 2214 | XP_011517019.2 |