Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCTCTAACCGTCGCTCCCTCCT[C/T]CCCTAGAAAGATGTTCTCGTCCGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600370 | ||||||||||||||||||||
Literature Links: |
SLC25A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC25A3 - solute carrier family 25 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002635.3 | 344 | Intron | NP_002626.1 | |||
NM_005888.3 | 344 | Intron | NP_005879.1 | |||
NM_213611.2 | 344 | UTR 5 | NP_998776.1 |
SNORA53 - small nucleolar RNA, H/ACA box 53 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |