Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCGTGTATTATTGTCCGAGTGG[C/G]CGGAACGGGAGCCAACATGGCAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607008 | ||||||||||||||||||||
Literature Links: |
ACADM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACADM - acyl-CoA dehydrogenase, C-4 to C-12 straight chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000016.5 | 425 | UTR 5 | NP_000007.1 | |||
NM_001127328.2 | 425 | UTR 5 | NP_001120800.1 | |||
NM_001286042.1 | 425 | UTR 5 | NP_001272971.1 | |||
NM_001286043.1 | 425 | UTR 5 | NP_001272972.1 | |||
NM_001286044.1 | 425 | UTR 5 | NP_001272973.1 |
SLC44A5 - solute carrier family 44 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130058.1 | 425 | Intron | NP_001123530.1 | |||
NM_001320283.1 | 425 | Intron | NP_001307212.1 | |||
NM_001320285.1 | 425 | Intron | NP_001307214.1 | |||
NM_001320287.1 | 425 | Intron | NP_001307216.1 | |||
NM_152697.4 | 425 | Intron | NP_689910.2 | |||
XM_006710445.3 | 425 | Intron | XP_006710508.1 | |||
XM_011540986.2 | 425 | Intron | XP_011539288.1 | |||
XM_011540987.2 | 425 | Intron | XP_011539289.1 | |||
XM_017000607.1 | 425 | Intron | XP_016856096.1 | |||
XM_017000608.1 | 425 | Intron | XP_016856097.1 | |||
XM_017000609.1 | 425 | Intron | XP_016856098.1 | |||
XM_017000610.1 | 425 | Intron | XP_016856099.1 | |||
XM_017000611.1 | 425 | Intron | XP_016856100.1 |