Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATTGAATATGGCCTCTTTTGT[T/G]GGATAATACCCATTATCATTCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607186 | ||||||||||||||||||||
Literature Links: |
SEC24D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEC24D - SEC24 homolog D, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318066.1 | 2388 | Missense Mutation | CAA,CCA | Q,P 979 | NP_001304995.1 | |
NM_014822.3 | 2388 | Missense Mutation | CAA,CCA | Q,P 978 | NP_055637.2 | |
XM_005263379.2 | 2388 | Missense Mutation | CAA,CCA | Q,P 979 | XP_005263436.1 | |
XM_017008875.1 | 2388 | Missense Mutation | CAA,CCA | Q,P 534 | XP_016864364.1 |