Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAATTAAGGAAGCAATTTTCTCC[C/T]TTTTACATGAACTGGATTTGACCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600410 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929319 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC101929319 - uncharacterized LOC101929319 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4773-1 - microRNA 4773-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4773-2 - microRNA 4773-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFAIP6 - TNF alpha induced protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007115.3 | Intron | NP_009046.2 |