Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605701 MIM: 616976 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRIM10 - tripartite motif containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006778.3 | 3248 | UTR 3 | NP_006769.2 | |||
NM_052828.2 | 3248 | UTR 3 | NP_439893.2 | |||
XM_011514221.1 | 3248 | UTR 3 | XP_011512523.1 | |||
XM_011514222.2 | 3248 | UTR 3 | XP_011512524.1 | |||
XM_011514223.2 | 3248 | UTR 3 | XP_011512525.1 | |||
XM_011514225.1 | 3248 | UTR 3 | XP_011512527.1 |
TRIM40 - tripartite motif containing 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |