Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAGGTTGGGGGCTAAGGTGGTA[A/G]TTTTGTGTTTTTTTGAGACGGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM254 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM254 - transmembrane protein 254 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270367.1 | Intron | NP_001257296.1 | ||||
NM_001270368.1 | Intron | NP_001257297.1 | ||||
NM_001270369.1 | Intron | NP_001257298.1 | ||||
NM_001270370.1 | Intron | NP_001257299.1 | ||||
NM_001270371.1 | Intron | NP_001257300.1 | ||||
NM_001270372.1 | Intron | NP_001257301.1 | ||||
NM_001270373.1 | Intron | NP_001257302.1 | ||||
NM_001270374.1 | Intron | NP_001257303.1 | ||||
NM_025125.3 | Intron | NP_079401.2 | ||||
XM_006717978.2 | Intron | XP_006718041.1 | ||||
XM_011540192.2 | Intron | XP_011538494.1 | ||||
XM_011540193.2 | Intron | XP_011538495.1 | ||||
XM_017016679.1 | Intron | XP_016872168.1 | ||||
XM_017016680.1 | Intron | XP_016872169.1 |
TMEM254-AS1 - TMEM254 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |