Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATAGAGACGAGGTCTCACTATTT[C/T]GCCCAAGCTGGTCTTAAACTTCTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611686 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CASD1 - CAS1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022900.4 | Intron | NP_075051.4 | ||||
XM_006716092.3 | Intron | XP_006716155.1 | ||||
XM_006716093.3 | Intron | XP_006716156.2 | ||||
XM_011516495.2 | Intron | XP_011514797.1 | ||||
XM_017012546.1 | Intron | XP_016868035.1 | ||||
XM_017012547.1 | Intron | XP_016868036.1 |
Set Membership: |
HapMap |