Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604942 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNPH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNPH - syntaphilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318234.1 | Intron | NP_001305163.1 | ||||
NM_014723.3 | Intron | NP_055538.2 | ||||
XM_005260889.3 | Intron | XP_005260946.1 | ||||
XM_011529402.2 | Intron | XP_011527704.1 | ||||
XM_011529403.2 | Intron | XP_011527705.1 | ||||
XM_011529404.2 | Intron | XP_011527706.1 |