Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608336 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6825 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR6825 - microRNA 6825 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPRA1 - transmembrane protein adipocyte associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136053.2 | Intron | NP_001129525.1 | ||||
NM_001142646.2 | Intron | NP_001136118.1 | ||||
XM_006713495.3 | Intron | XP_006713558.1 | ||||
XM_006713496.2 | Intron | XP_006713559.1 | ||||
XM_006713497.3 | Intron | XP_006713560.1 | ||||
XM_011512422.1 | Intron | XP_011510724.1 | ||||
XM_017005707.1 | Intron | XP_016861196.1 | ||||
XM_017005708.1 | Intron | XP_016861197.1 | ||||
XM_017005709.1 | Intron | XP_016861198.1 | ||||
XM_017005710.1 | Intron | XP_016861199.1 |