Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613747 MIM: 602852 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF24 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIF24 - kinesin family member 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194313.2 | Intron | NP_919289.2 | ||||
XM_005251451.2 | Intron | XP_005251508.1 | ||||
XM_006716764.2 | Intron | XP_006716827.1 | ||||
XM_011517861.1 | Intron | XP_011516163.1 | ||||
XM_011517863.2 | Intron | XP_011516165.1 | ||||
XM_017014697.1 | Intron | XP_016870186.1 | ||||
XM_017014698.1 | Intron | XP_016870187.1 |
NUDT2 - nudix hydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161.4 | Intron | NP_001152.1 | ||||
NM_001244390.1 | Intron | NP_001231319.1 | ||||
NM_147172.2 | Intron | NP_671701.1 | ||||
NM_147173.2 | Intron | NP_671702.1 |
Set Membership: |
HapMap |