Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610078 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MORC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MORC3 - MORC family CW-type zinc finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320445.1 | Intron | NP_001307374.1 | ||||
NM_001320446.1 | Intron | NP_001307375.1 | ||||
NM_015358.2 | Intron | NP_056173.1 | ||||
XM_011529518.2 | Intron | XP_011527820.1 | ||||
XM_017028311.1 | Intron | XP_016883800.1 | ||||
XM_017028312.1 | Intron | XP_016883801.1 |