Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605134 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PITPNC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PITPNC1 - phosphatidylinositol transfer protein, cytoplasmic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012417.3 | 583 | UTR 5 | NP_036549.2 | |||
NM_181671.2 | 583 | UTR 5 | NP_858057.1 | |||
XM_005257216.2 | 583 | Intron | XP_005257273.1 | |||
XM_017024442.1 | 583 | Intron | XP_016879931.1 | |||
XM_017024443.1 | 583 | Intron | XP_016879932.1 | |||
XM_017024444.1 | 583 | Intron | XP_016879933.1 | |||
XM_017024445.1 | 583 | Intron | XP_016879934.1 |
Set Membership: |
HapMap |