Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604392 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AIPL1 - aryl hydrocarbon receptor interacting protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033054.2 | 2569 | UTR 3 | NP_001028226.1 | |||
NM_001033055.2 | 2569 | UTR 3 | NP_001028227.1 | |||
NM_001285399.2 | 2569 | UTR 3 | NP_001272328.1 | |||
NM_001285400.2 | 2569 | UTR 3 | NP_001272329.1 | |||
NM_001285401.2 | 2569 | UTR 3 | NP_001272330.1 | |||
NM_001285402.1 | 2569 | UTR 3 | NP_001272331.1 | |||
NM_001285403.2 | 2569 | Intron | NP_001272332.1 | |||
NM_014336.4 | 2569 | UTR 3 | NP_055151.3 |
Set Membership: |
HapMap |