Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601097 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PMP22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PMP22 - peripheral myelin protein 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000304.3 | 1793 | UTR 3 | NP_000295.1 | |||
NM_001281455.1 | 1793 | UTR 3 | NP_001268384.1 | |||
NM_001281456.1 | 1793 | UTR 3 | NP_001268385.1 | |||
NM_153321.2 | 1793 | UTR 3 | NP_696996.1 | |||
NM_153322.2 | 1793 | UTR 3 | NP_696997.1 | |||
XM_017024775.1 | 1793 | Intron | XP_016880264.1 | |||
XM_017024776.1 | 1793 | Intron | XP_016880265.1 |