Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCCGGCCCCGGCGGCCGCAAGC[A/C]GGAGCGGCGGAACGTGTTTCCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600542 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928438 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928438 - uncharacterized LOC101928438 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR4A3 - nuclear receptor subfamily 4 group A member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006981.3 | Intron | NP_008912.2 | ||||
NM_173199.2 | Intron | NP_775291.1 | ||||
NM_173200.2 | Intron | NP_775292.1 | ||||
XM_017015162.1 | Intron | XP_016870651.1 |
Set Membership: |
HapMap |