Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605729 MIM: 138600 MIM: 138610 | ||||||||||||||||||||
Literature Links: |
AKNA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKNA - AT-hook transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORM1 - orosomucoid 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000607.2 | 503 | Missense Mutation | GCT,GGT | A,G 131 | NP_000598.2 |
ORM2 - orosomucoid 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |