Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGGGAGGATAGATCTGTTAAAG[A/T]GAACATGGAGCTGAAATCACATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605734 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEFF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEFF2 - transmembrane protein with EGF like and two follistatin like domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305134.1 | Intron | NP_001292063.1 | ||||
NM_001305145.1 | Intron | NP_001292074.1 | ||||
NM_016192.3 | Intron | NP_057276.2 | ||||
XM_011510890.2 | Intron | XP_011509192.1 | ||||
XM_017003739.1 | Intron | XP_016859228.1 | ||||
XM_017003740.1 | Intron | XP_016859229.1 |
Set Membership: |
HapMap |