Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCTATTTGGTTGCCCAATGATT[C/T]CCTAGGAGAAAAGAAGTTGAATATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300922 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHIC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHIC1 - cysteine rich hydrophobic domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039840.2 | Intron | NP_001034929.2 | ||||
NM_001300884.1 | Intron | NP_001287813.1 | ||||
XM_017029582.1 | Intron | XP_016885071.1 | ||||
XM_017029583.1 | Intron | XP_016885072.1 | ||||
XM_017029584.1 | Intron | XP_016885073.1 |