Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCAGGGACAGAGTTGACATCAC[C/T]GGGCATTTTAGCCAAAAAGCAGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611527 MIM: 611789 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC9B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC9B1 - solute carrier family 9 member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100874.2 | 2719 | Intron | NP_001094344.1 | |||
NM_139173.3 | 2719 | Intron | NP_631912.2 | |||
XM_006714093.3 | 2719 | Intron | XP_006714156.1 | |||
XM_011531622.1 | 2719 | Intron | XP_011529924.1 | |||
XM_011531623.1 | 2719 | Intron | XP_011529925.1 |
SLC9B2 - solute carrier family 9 member B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300754.1 | 2719 | Intron | NP_001287683.1 | |||
NM_001300756.1 | 2719 | Intron | NP_001287685.1 | |||
NM_178833.5 | 2719 | Intron | NP_849155.2 | |||
XM_006714085.2 | 2719 | UTR 3 | XP_006714148.1 | |||
XM_006714086.3 | 2719 | Intron | XP_006714149.1 |