Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTCCTCAGGCCCTAGTCCAGAG[A/G]TGACGTGTCTGTGGGAATGTTTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610196 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELMOD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ELMOD2 - ELMO domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153702.3 | Intron | NP_714913.1 | ||||
XM_005262885.3 | Intron | XP_005262942.1 | ||||
XM_005262886.2 | Intron | XP_005262943.1 | ||||
XM_011531818.2 | Intron | XP_011530120.1 | ||||
XM_011531819.2 | Intron | XP_011530121.1 |
Set Membership: |
HapMap |