Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGGGCCTGATTTGGCTCACTC[G/T]TGTGGATTCCTGGCACCCAGAAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607870 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC5B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UNC5B - unc-5 netrin receptor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244889.1 | Intron | NP_001231818.1 | ||||
NM_170744.4 | Intron | NP_734465.2 | ||||
XM_011539453.1 | Intron | XP_011537755.1 | ||||
XM_017015834.1 | Intron | XP_016871323.1 | ||||
XM_017015835.1 | Intron | XP_016871324.1 |
UNC5B-AS1 - UNC5B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |