Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTAAACTATCTCTGCAAATTTT[C/T]TTGACACTCCTCTATTCAAAAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610371 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC2A7 - solute carrier family 2 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207420.2 | Intron | NP_997303.2 | ||||
XM_011540824.2 | Intron | XP_011539126.1 | ||||
XM_011540825.2 | Intron | XP_011539127.1 |