Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612811 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRFN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRFN5 - leucine rich repeat and fibronectin type III domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152447.3 | 755 | Intron | NP_689660.2 | |||
XM_005267369.3 | 755 | UTR 5 | XP_005267426.1 | |||
XM_017021048.1 | 755 | Intron | XP_016876537.1 | |||
XM_017021049.1 | 755 | UTR 5 | XP_016876538.1 | |||
XM_017021050.1 | 755 | UTR 5 | XP_016876539.1 |
Set Membership: |
HapMap |