Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604911 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CNOT4 - CCR4-NOT transcription complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008225.2 | Intron | NP_001008226.1 | ||||
NM_001190847.1 | Intron | NP_001177776.1 | ||||
NM_001190848.1 | Intron | NP_001177777.1 | ||||
NM_001190849.1 | Intron | NP_001177778.1 | ||||
NM_001190850.1 | Intron | NP_001177779.1 | ||||
NM_013316.3 | Intron | NP_037448.2 | ||||
XM_017012235.1 | Intron | XP_016867724.1 |
Set Membership: |
HapMap Validated |