Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCGGAATAACACAAGAGCAACA[A/G]TAATAATGTACTCATTTACTAAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604055 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IDI1 - isopentenyl-diphosphate delta isomerase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317955.1 | Intron | NP_001304884.1 | ||||
NM_001317956.1 | Intron | NP_001304885.1 | ||||
NM_001317957.1 | Intron | NP_001304886.1 | ||||
NM_004508.3 | Intron | NP_004499.2 | ||||
XM_011519457.1 | Intron | XP_011517759.1 | ||||
XM_017016191.1 | Intron | XP_016871680.1 |
WDR37 - WD repeat domain 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014023.3 | Intron | NP_054742.2 |