Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCTAGGCTGAAGGTGGTGGCTG[A/G]GGCTGTCCACCTGCCCAGCCTCCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610994 MIM: 602995 | ||||||||||||||||||||
Literature Links: |
TMEM189 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM189 - transmembrane protein 189 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162505.1 | 1452 | UTR 3 | NP_001155977.1 | |||
NM_199129.2 | 1452 | UTR 3 | NP_954580.1 |
TMEM189-UBE2V1 - TMEM189-UBE2V1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199203.2 | 1452 | Intron | NP_954673.1 |
UBE2V1 - ubiquitin conjugating enzyme E2 V1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |