Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107910 MIM: 608603 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP19A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP19A1 - cytochrome P450 family 19 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GLDN - gliomedin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181789.2 | Intron | NP_861454.2 | ||||
XM_011521501.2 | Intron | XP_011519803.1 | ||||
XM_017022121.1 | Intron | XP_016877610.1 | ||||
XM_017022122.1 | Intron | XP_016877611.1 | ||||
XM_017022123.1 | Intron | XP_016877612.1 | ||||
XM_017022124.1 | Intron | XP_016877613.1 | ||||
XM_017022125.1 | Intron | XP_016877614.1 | ||||
XM_017022126.1 | Intron | XP_016877615.1 |
Set Membership: |
HapMap |