Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TTC13 - tetratricopeptide repeat domain 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122835.2 | Intron | NP_001116307.2 | ||||
NM_024525.4 | Intron | NP_078801.3 | ||||
XM_005273260.1 | Intron | XP_005273317.1 | ||||
XM_005273261.2 | Intron | XP_005273318.1 | ||||
XM_005273262.1 | Intron | XP_005273319.1 | ||||
XM_005273264.2 | Intron | XP_005273321.1 | ||||
XM_006711814.2 | Intron | XP_006711877.1 | ||||
XM_006711815.2 | Intron | XP_006711878.1 | ||||
XM_011544275.2 | Intron | XP_011542577.1 | ||||
XM_011544276.2 | Intron | XP_011542578.1 | ||||
XM_017002321.1 | Intron | XP_016857810.1 | ||||
XM_017002322.1 | Intron | XP_016857811.1 | ||||
XM_017002323.1 | Intron | XP_016857812.1 | ||||
XM_017002324.1 | Intron | XP_016857813.1 | ||||
XM_017002325.1 | Intron | XP_016857814.1 | ||||
XM_017002326.1 | Intron | XP_016857815.1 |