Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf33 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C19orf33 - chromosome 19 open reading frame 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317801.1 | 2218 | Intron | NP_001304730.1 | |||
NM_033520.2 | 2218 | Intron | NP_277055.1 |
YIF1B - Yip1 interacting factor homolog B, membrane trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039671.2 | 2218 | UTR 3 | NP_001034760.1 | |||
NM_001039672.2 | 2218 | UTR 3 | NP_001034761.1 | |||
NM_001039673.2 | 2218 | UTR 3 | NP_001034762.1 | |||
NM_001145461.1 | 2218 | UTR 3 | NP_001138933.1 | |||
NM_001145462.1 | 2218 | UTR 3 | NP_001138934.1 | |||
NM_001145463.1 | 2218 | UTR 3 | NP_001138935.1 | |||
XM_005259385.3 | 2218 | Intron | XP_005259442.1 | |||
XM_017027449.1 | 2218 | Intron | XP_016882938.1 | |||
XM_017027450.1 | 2218 | Intron | XP_016882939.1 | |||
XM_017027451.1 | 2218 | Intron | XP_016882940.1 |
Set Membership: |
HapMap |