Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAACAAGGAAGTAATTGGGTCAT[C/G]TTGAATCAACAAGGAAGTAATTAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611232 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLDN12 - claudin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185072.2 | Intron | NP_001172001.1 | ||||
NM_001185073.2 | Intron | NP_001172002.1 | ||||
NM_012129.4 | Intron | NP_036261.1 |
LOC102723899 - pituitary tumor-transforming gene 1 protein-interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011516764.2 | Intron | XP_011515066.1 | ||||
XM_017012889.1 | Intron | XP_016868378.1 | ||||
XM_017012890.1 | Intron | XP_016868379.1 | ||||
XM_017012891.1 | Intron | XP_016868380.1 | ||||
XM_017012892.1 | Intron | XP_016868381.1 | ||||
XM_017012893.1 | Intron | XP_016868382.1 | ||||
XM_017012894.1 | Intron | XP_016868383.1 |